Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pycrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025412.2 | NP_079688.2 | 6 | 814 | ||
GenBank mRNA | AK002912.1 | - | 6 | 745 | ||
AK013214.1 | - | 6 | 739 | |||
BC026536.1 | - | 6 | 735 | 4987954 | ||
BF721406.1 | - | 1 | 538 |