Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033132.3 | NP_001028304.1 | 3 | 164 | ||
NM_001168592.1 | NP_001162064.1 | 2 | 152 | |||
GenBank mRNA | AK004316.1 | - | 3 | 138 | ||
AK008220.1 | - | 3 | 146 | |||
AK009796.1 | - | 3 | 159 | |||
AK011226.1 | - | 3 | 149 | |||
AK018711.1 | - | 3 | 138 | |||
AK019351.1 | - | 3 | 140 | |||
AK019432.1 | - | 3 | 141 | |||
AK020234.1 | - | 3 | 137 | |||
AK034223.1 | - | 1 | 3412 | |||
AK045407.1 | - | 2638 | ||||
BB577089.1 | - | 2 | 165 | |||
BC132261.1 | - | 3 | 151 | 40130538 | ||
BC132265.1 | - | 3 | 151 | 40130542 | ||
BC145360.1 | - | 3 | 151 | 9053918 | ||
BC145361.1 | - | 2 | 139 | 9053919 | ||
CK621880.1 | - | 2 | 115 |