Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104530.1 | NP_001098000.1 | 3 | 398 | ||
NM_001104556.1 | NP_001098026.1 | 2 | 396 | |||
NM_001104557.1 | NP_001098027.1 | 2 | 228 | |||
NM_025426.3 | NP_079702.3 | 3 | 566 | |||
GenBank mRNA | AK004354.1 | - | 3 | 531 | ||
AK010357.1 | - | 3 | 394 | |||
AK012799.1 | - | 3 | 546 | |||
AK144918.1 | - | 2 | 354 | |||
AK147739.1 | - | 3 | 556 | |||
AK167407.1 | - | 3 | 526 | |||
BE333513.1 | - | 2 | 326 | |||
C88719.1 | - | 2 | 201 | |||
CK030901.1 | - | 3 | 398 | |||
CK788351.1 | - | 218 |
Gene Symbol: 2310031A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK009549.1 | - | 2 | 638 |