Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpinb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025429.2 | NP_079705.2 | 7 | 853 | ||
GenBank mRNA | AF426024.1 | - | 844 | |||
AK003930.1 | - | 7 | 848 | |||
AK008914.1 | - | 7 | 848 | |||
AK076129.1 | - | 7 | 848 | |||
AK160167.1 | - | 7 | 849 | |||
BC011140.1 | - | 832 | 4207111 | |||
BC104333.2 | - | 6 | 820 | 40045808 |