Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrps28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025434.3 | NP_079710.3 | 2 | 310 | ||
GenBank mRNA | AK005229.1 | - | 2 | 310 | ||
AK011036.1 | - | 2 | 271 | |||
AK088314.1 | - | 2 | 258 | |||
BC028530.1 | - | 2 | 276 | 1381263 |