Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1ax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025437.4 | NP_079713.2 | 2, 3 | 309 | ||
GenBank mRNA | AK005185.1 | - | 2, 3 | 297 | ||
AK007301.1 | - | 2, 3 | 260 | |||
AK012233.1 | - | 2, 3 | 289 | |||
AK012464.1 | - | 2, 3 | 210 | |||
AK012714.1 | - | 2, 3 | 292 | |||
AK021283.1 | - | 2, 3 | 290 | |||
AK030774.1 | - | 313 | ||||
AK031113.1 | - | 321 | ||||
AK077710.1 | - | 2, 3 | 258 | |||
AK090053.1 | - | 275 | ||||
AK134827.1 | - | 234 | ||||
AK134969.1 | - | 234 | ||||
AK165574.1 | - | 275 | ||||
AK166629.1 | - | 2, 3 | 312 | |||
AK166852.1 | - | 2, 3 | 310 | |||
AK187388.1 | - | 1, 2 | 49 | |||
AK207164.1 | - | 1, 2 | 34 | |||
AK210743.1 | - | 1, 2 | 23 | |||
AK217305.1 | - | 1, 2 | 71 | |||
BC027284.1 | - | 2, 3 | 246 | 3156134 |