Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1ax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025437.4 | NP_079713.2 | 4, 5 | 468 | ||
GenBank mRNA | AK005185.1 | - | 4, 5 | 456 | ||
AK007301.1 | - | 4, 5 | 419 | |||
AK012233.1 | - | 4, 5 | 448 | |||
AK012714.1 | - | 4, 5 | 451 | |||
AK021283.1 | - | 4, 5 | 449 | |||
AK030774.1 | - | 472 | ||||
AK031113.1 | - | 480 | ||||
AK077710.1 | - | 4, 5 | 417 | |||
AK090053.1 | - | 434 | ||||
AK134827.1 | - | 393 | ||||
AK134969.1 | - | 393 | ||||
AK165574.1 | - | 434 | ||||
AK166629.1 | - | 4, 5 | 471 | |||
AK166852.1 | - | 4, 5 | 469 | |||
AK187388.1 | - | 3, 4 | 209 | |||
AK207164.1 | - | 3, 4 | 193 | |||
AK217305.1 | - | 3, 4 | 230 | |||
BC027284.1 | - | 4, 5 | 405 | 3156134 |