Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023179.3 | NP_075668.1 | 1 | 183 | ||
GenBank mRNA | AB076405.1 | - | 1 | 238 | ||
AK005109.1 | - | 1 | 64 | |||
AK011945.1 | - | 1 | 26 | |||
BC020190.1 | - | 1 | 179 | 4505711 | ||
BC049587.1 | - | 1 | 48 | |||
BC062380.1 | - | 1 | 171 | 6468413 | ||
BU558667.1 | - | 1 | 186 |