Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dimt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025447.4 | NP_079723.1 | 8 | 697 | ||
GenBank mRNA | AK007759.1 | - | 8 | 648 | ||
AK011544.1 | - | 8 | 654 | |||
AK015145.1 | - | 8 | 647 | |||
AK078186.1 | - | 6 | 1491 | |||
AK078401.1 | - | 6 | 1513 | |||
AK146188.1 | - | 8 | 668 | |||
AK161333.1 | - | 8 | 649 | |||
BC019799.1 | - | 8 | 670 | 5053500 |