Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025461.5 | NP_079737.1 | 1 | 185 | ||
GenBank mRNA | AK007450.1 | - | 1 | 162 | ||
AK007562.1 | - | 1 | 174 | |||
AK008486.1 | - | 1 | 150 | |||
AK040003.1 | - | 1 | 187 | |||
AK054491.1 | - | 1 | 161 | |||
AK079975.1 | - | 132 | ||||
AK082374.1 | - | 1 | 186 | |||
AK084315.1 | - | 1 | 182 | |||
AK087666.1 | - | 186 | ||||
AK088794.1 | - | 186 | ||||
BC005675.1 | - | 149 | 3153356 | |||
BC100533.1 | - | 116 | 30134701 |