Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025461.5 | NP_079737.1 | 1, 2 | 211 | ||
GenBank mRNA | AK007450.1 | - | 1, 2 | 188 | ||
AK007562.1 | - | 1, 2 | 200 | |||
AK008486.1 | - | 1, 2 | 176 | |||
AK040003.1 | - | 1, 2 | 213 | |||
AK054491.1 | - | 1, 2 | 187 | |||
AK079975.1 | - | 158 | ||||
AK082374.1 | - | 1, 2 | 212 | |||
AK087666.1 | - | 212 | ||||
AK088794.1 | - | 212 | ||||
BC005675.1 | - | 175 | 3153356 | |||
BC100533.1 | - | 142 | 30134701 |