Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025461.5 | NP_079737.1 | 2, 3 | 284 | ||
GenBank mRNA | AK007450.1 | - | 2, 3 | 261 | ||
AK007562.1 | - | 2, 3 | 273 | |||
AK007870.1 | - | 2, 3 | 157 | |||
AK008486.1 | - | 2, 3 | 249 | |||
AK040003.1 | - | 2, 3 | 286 | |||
AK079975.1 | - | 231 | ||||
AK082374.1 | - | 2, 3 | 285 | |||
AK087666.1 | - | 285 | ||||
AK088794.1 | - | 285 | ||||
BC005675.1 | - | 248 | 3153356 | |||
BC100533.1 | - | 215 | 30134701 |