Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cox20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025511.2 | NP_079787.1 | 2 | 102 | ||
GenBank mRNA | AK009178.1 | - | 2 | 96 | ||
BC055277.1 | - | 2 | 85 | 5714061 | ||
BC059717.1 | - | 2 | 100 | 6543216 | ||
BY779933.1 | - | 2 | 102 |
Gene Symbol: AI503316 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK153188.1 | - | 1 | 441 | ||
BC033430.1 | - | 2 | 1784 |