Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025527.3 | NP_079803.2 | 2, 3 | 183 | ||
GenBank mRNA | AK004118.1 | - | 2, 3 | 185 | ||
AK009412.1 | - | 2, 3 | 147 | |||
AK010760.1 | - | 2, 3 | 182 | |||
AK152322.1 | - | 2, 3 | 162 | |||
AK162107.1 | - | 2, 3 | 153 | |||
BC051932.1 | - | 2, 3 | 150 | 5710637 |