Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srp19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025527.3 | NP_079803.2 | 4 | 303 | ||
GenBank mRNA | AK004118.1 | - | 4 | 305 | ||
AK009412.1 | - | 4 | 267 | |||
AK010760.1 | - | 4 | 302 | |||
AK037009.1 | - | 3 | 2773 | |||
AK135508.1 | - | 5 | 430 | |||
AK152322.1 | - | 4 | 282 | |||
AK162107.1 | - | 4 | 273 | |||
BC051932.1 | - | 4 | 270 | 5710637 |