Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133435.1 | NP_597679.1 | 3 | 337 | ||
GenBank mRNA | AF260925.1 | - | 3 | 490 | ||
AK011137.1 | - | 3 | 359 | |||
AK012194.1 | - | 3 | 571 | |||
AK017631.1 | - | 347 | ||||
AK152407.1 | - | 397 | ||||
AY679721.1 | - | 3 | 337 | |||
BC138500.1 | - | 3 | 405 | 8861520 | ||
BC138502.1 | - | 3 | 405 | 8861522 |