Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133435.1 | NP_597679.1 | 4, 5 | 537 | ||
GenBank mRNA | AF260925.1 | - | 4, 5 | 690 | ||
AK011137.1 | - | 4, 5 | 559 | |||
AK012194.1 | - | 4, 5 | 771 | |||
AY679721.1 | - | 4, 5 | 537 | |||
BC138500.1 | - | 4, 5 | 605 | 8861520 | ||
BC138502.1 | - | 4, 5 | 605 | 8861522 |