Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nmnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133435.1 | NP_597679.1 | 5 | 579 | ||
GenBank mRNA | AF260925.1 | - | 5 | 732 | ||
AK012194.1 | - | 5 | 813 | |||
AK152407.1 | - | 1067 | ||||
AY679721.1 | - | 5 | 579 | |||
BC138500.1 | - | 5 | 647 | 8861520 | ||
BC138502.1 | - | 5 | 647 | 8861522 |
Gene Symbol: Erdr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC038133.1 | - | 179 |