Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025575.3 | NP_079851.2 | 3, 4 | 462 | ||
GenBank mRNA | AK003071.1 | - | 3, 4 | 455 | ||
AK005249.1 | - | 3, 4 | 365 | |||
AK005510.1 | - | 3, 4 | 399 | |||
AK007882.1 | - | 3, 4 | 461 | |||
AK009684.1 | - | 3, 4 | 426 | |||
AK011753.1 | - | 3, 4 | 458 | |||
AK013220.1 | - | 3, 4 | 435 | |||
AK017524.1 | - | 2, 3 | 294 | |||
AK032098.1 | - | 2, 3 | 297 | |||
AK075770.1 | - | 3, 4 | 427 | |||
AK148305.1 | - | 3, 4 | 400 | |||
BC038166.1 | - | 1, 2 | 408 | |||
BC053943.1 | - | 2, 3 | 296 | |||
BC054055.1 | - | 1, 2 | 580 | |||
BC061084.1 | - | 2, 3 | 336 | 6596164 |