Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sys1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025575.3 | NP_079851.2 | 4 | 648 | ||
GenBank mRNA | AK003071.1 | - | 4 | 641 | ||
AK005249.1 | - | 4 | 551 | |||
AK007882.1 | - | 4 | 647 | |||
AK009684.1 | - | 4 | 612 | |||
AK011753.1 | - | 4 | 644 | |||
AK013220.1 | - | 4 | 621 | |||
AK017524.1 | - | 3 | 480 | |||
AK032098.1 | - | 3 | 483 | |||
AK075770.1 | - | 4 | 613 | |||
AK148305.1 | - | 4 | 586 | |||
BC038166.1 | - | 1 | 222 | |||
BC053943.1 | - | 3 | 482 | |||
BC054055.1 | - | 1 | 394 | |||
BC061084.1 | - | 3 | 522 | 6596164 |