Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025576.2 | NP_079852.1 | 2 | 504 | ||
GenBank mRNA | AK003210.1 | - | 1 | 150 | ||
AK012674.1 | - | 2 | 494 | |||
AK014691.1 | - | 2 | 498 | |||
AK151972.1 | - | 2 | 505 | |||
BC026750.1 | - | 2 | 483 | 5135841 |