Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109743.1 | NP_001103213.1 | 2 | 121 | ||
GenBank mRNA | AB358976.1 | - | 2 | 121 |