Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163007.1 | NP_001156479.1 | 23 | 3186 | ||
NM_001163008.1 | NP_001156480.1 | 22 | 3099 | |||
NM_001163009.1 | NP_001156481.1 | 23 | 3186 | |||
NM_026910.1 | NP_081186.1 | 24 | 3210 | |||
GenBank mRNA | AK088459.1 | - | 4 | 468 | ||
AK122306.1 | - | 3 | 967 | |||
BC050866.1 | - | 23 | 3203 | |||
BC050868.1 | - | 23 | 3203 | |||
BC137799.1 | - | 24 | 2861 | 8860818 | ||
BC151003.1 | - | 23 | 2837 | 9087916 | ||
BC158011.1 | - | 22 | 2750 | 9007319 | ||
BC158059.1 | - | 23 | 2837 | 9007359 | ||
CD348964.1 | - | 8 | 784 |