Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026844.3 | NP_081120.2 | 3, 4 | 253 | ||
GenBank mRNA | AK002429.1 | - | 3, 4 | 253 | ||
AK004189.1 | - | 4, 5 | 445 | |||
AK009995.1 | - | 3, 4 | 284 | |||
AK010073.1 | - | 3, 4 | 284 | |||
AK013475.1 | - | 3, 4 | 250 | |||
BC027549.1 | - | 3, 4 | 223 | 1548292 |