Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntpcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025636.5 | NP_079912.1 | 3 | 391 | ||
GenBank mRNA | AK006433.1 | - | 3 | 299 | ||
AK010229.1 | - | 3 | 366 | |||
AK010823.1 | - | 3 | 392 | |||
AK019321.1 | - | 3 | 371 | |||
AK019421.1 | - | 3 | 362 | |||
AK086714.1 | - | 3 | 357 | |||
BC025168.1 | - | 3 | 299 | 4223622 |