Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Farsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025648.3 | NP_079924.2 | 8, 9 | 972 | ||
GenBank mRNA | AK002577.1 | - | 8, 9 | 948 | ||
AK016506.1 | - | 8, 9 | 968 | |||
AK031697.1 | - | 8, 9 | 950 | |||
AK154629.1 | - | 8, 9 | 970 | |||
AK171872.1 | - | 8, 9 | 852 | |||
BC006862.1 | - | 8, 9 | 943 | 3595108 | ||
BC013533.1 | - | 8, 9 | 941 | 4238911 |