Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025652.3 | NP_079928.2 | 5 | 655 | ||
GenBank mRNA | AF391799.1 | - | 5 | 639 | ||
AK008247.2 | - | 5 | 623 | |||
AK011845.1 | - | 5 | 604 | |||
AK012797.1 | - | 5 | 602 | |||
AK017234.1 | - | 5 | 549 | |||
AK143627.1 | - | 5 | 607 | |||
BC032292.1 | - | 4 | 310 | 5374268 | ||
BC057966.1 | - | 4 | 867 | |||
BC064891.1 | - | 3 | 326 |
Gene Symbol: Mtif3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256100.1 | NP_001243029.1 | 5 | 3893 | ||
NM_001256101.1 | NP_001243030.1 | 5 | 3882 | |||
NM_001256102.1 | NP_001243031.1 | 4 | 3722 | |||
NM_029581.4 | NP_083857.1 | 4 | 3733 | |||
GenBank mRNA | BC052439.1 | - | 4 | 3686 | 6406303 |