Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081172.2 | NP_001074641.1 | 16 | 3356 | ||
GenBank mRNA | AK122409.1 | - | 13 | 2985 | ||
AK133351.1 | - | 3 | 1795 | |||
BC031840.1 | - | 6 | 2216 | |||
BC046469.1 | - | 2 | 1132 | |||
BC051097.1 | - | 6 | 2216 | |||
BC158060.1 | - | 16 | 3320 | 9007360 |