Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081172.2 | NP_001074641.1 | 16 | 4541 | ||
GenBank mRNA | AK122409.1 | - | 13 | 4167 | ||
AK133351.1 | - | 3 | 2980 | |||
BC031840.1 | - | 6 | 3401 | |||
BC046469.1 | - | 2 | 2317 | |||
BC051097.1 | - | 6 | 3401 | |||
BC158060.1 | - | 16 | 4505 | 9007360 | ||
BG070589.2 | - | 1 | 268 | |||
BM213356.2 | - | 1 | 268 |