Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryzl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026994.1 | NP_081270.1 | 9 | 975 | ||
NM_133679.2 | NP_598440.1 | 10 | 1020 | |||
GenBank mRNA | AK051915.1 | - | 1 | 2411 | ||
AK145827.1 | - | 10 | 844 | |||
AK170997.1 | - | 2 | 136 | |||
BC010479.1 | - | 10 | 817 | 3968727 | ||
BC019387.1 | - | 9 | 784 | 4164847 |