Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025664.5 | NP_079940.2 | 17 | 1840 | ||
GenBank mRNA | AK004449.1 | - | 211 | |||
AK012520.1 | - | 12 | 1132 | |||
AK139802.1 | - | 17 | 1874 | |||
AK149739.1 | - | 17 | 1853 | |||
AK149849.1 | - | 17 | 1894 | |||
AK155830.1 | - | 1875 | ||||
AK157517.1 | - | 17 | 1856 | |||
AK170339.1 | - | 1856 | ||||
AK189192.1 | - | 3 | 175 | |||
BC014814.1 | - | 17 | 1840 | 3257422 |