Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025664.5 | NP_079940.2 | 18 | 1928 | ||
GenBank mRNA | AK004449.1 | - | 299 | |||
AK012520.1 | - | 13 | 1220 | |||
AK139802.1 | - | 18 | 1963 | |||
AK149739.1 | - | 18 | 1941 | |||
AK149849.1 | - | 18 | 1982 | |||
AK155830.1 | - | 1963 | ||||
AK157517.1 | - | 18 | 1944 | |||
AK170339.1 | - | 1944 | ||||
AK189192.1 | - | 4 | 263 | |||
BC014814.1 | - | 18 | 1928 | 3257422 |