Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spcs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025668.3 | NP_079944.1 | 2 | 132 | ||
GenBank mRNA | AK014310.1 | - | 2 | 146 | ||
AK017504.1 | - | 2 | 137 | |||
AK077510.1 | - | 2 | 142 | |||
AK134578.1 | - | 2 | 133 | |||
AK168173.1 | - | 2 | 137 | |||
BC050150.1 | - | 2 | 142 | 30026746 |