Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025673.2 | NP_079949.1 | 4 | 901 | ||
GenBank mRNA | AB271918.1 | - | 2 | 654 | ||
AK005996.1 | - | 4 | 948 | |||
AK015543.1 | - | 4 | 945 | |||
AK028409.1 | - | 4 | 912 | |||
AK077446.1 | - | 4 | 966 | |||
AK133763.1 | - | 4 | 949 | |||
AK136194.1 | - | 4 | 526 | |||
AK139545.1 | - | 4 | 948 | |||
BC027333.1 | - | 1 | 1097 | |||
BC031445.1 | - | 5 | 811 | 4482612 | ||
BC039909.1 | - | 4 | 898 | |||
BC051035.1 | - | 4 | 901 | 2655316 |