Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpgs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004361.2 | NP_001004361.2 | 6 | 895 | ||
NM_001142697.1 | NP_001136169.1 | 6 | 641 | |||
NM_001142698.1 | NP_001136170.1 | 6 | 895 | |||
GenBank mRNA | AK141632.1 | - | 6 | 712 | ||
AK160220.1 | - | 6 | 691 | |||
AK178089.1 | - | 1 | 47 | |||
BC060253.1 | - | 4 | 447 | |||
BC080777.1 | - | 6 | 651 | 30439612 | ||
DV655125.1 | - | 6 | 641 |