Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sltm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025690.3 | NP_079966.2 | 21 | 3076 | ||
NM_026337.1 | NP_080613.1 | 21 | 3114 | |||
GenBank mRNA | AF462146.1 | - | 21 | 3076 | ||
AK134756.1 | - | 12 | 1755 | |||
BC019992.1 | - | 10 | 1446 | |||
BC139202.1 | - | 21 | 3066 | 8862224 | ||
BC145577.1 | - | 20 | 2885 | |||
CN527065.1 | - | 3 | 250 |