Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem41a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025693.4 | NP_079969.1 | 4, 5 | 639 | ||
GenBank mRNA | AK007685.1 | - | 4, 5 | 634 | ||
AK013518.1 | - | 3, 4 | 464 | |||
AK160292.1 | - | 4, 5 | 560 | |||
BC011512.1 | - | 4, 5 | 619 | |||
BC019770.1 | - | 4, 5 | 594 | 5124769 |