Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msantd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145924.1 | NP_001139396.1 | 3 | 834 | ||
NM_001145925.1 | NP_001139397.1 | 3 | 695 | |||
NM_028137.3 | NP_082413.2 | 3 | 713 | |||
GenBank mRNA | AK011381.1 | - | 3 | 836 | ||
AK013199.1 | - | 3 | 714 | |||
AK017795.1 | - | 3 | 838 | |||
AK075956.1 | - | 3 | 707 | |||
BC028651.1 | - | 2 | 236 | |||
BC052696.1 | - | 3 | 756 | 5715147 |