Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025701.4 | NP_079977.3 | 2 | 220 | ||
GenBank mRNA | AK003633.1 | - | 2 | 188 | ||
AK007989.1 | - | 2 | 187 | |||
AK076212.1 | - | 2 | 221 | |||
AK078451.1 | - | 2 | 186 | |||
AK171833.1 | - | 1 | 2600 | |||
BC028946.1 | - | 1 | 680 | |||
BC049179.1 | - | 2 | 178 | 6313540 |