Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrfs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025710.2 | NP_079986.1 | 2 | 672 | ||
GenBank mRNA | AK003966.1 | - | 2 | 635 | ||
AK012180.1 | - | 2 | 623 | |||
AK014470.1 | - | 2 | 675 | |||
AK152391.1 | - | 2 | 684 | |||
AK153139.1 | - | 2 | 659 | |||
AK180820.1 | - | 1 | 208 | |||
AK191429.1 | - | 1 | 181 | |||
AK197173.1 | - | 1 | 155 | |||
AK210928.1 | - | 1 | 157 | |||
AK212564.1 | - | 1 | 169 | |||
AK216624.1 | - | 1 | 169 | |||
AK217705.1 | - | 1 | 169 | |||
BC019934.1 | - | 2 | 679 | 5249225 | ||
BC047125.1 | - | 2 | 679 |