Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm8439 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101603.1 | NP_001095073.1 | 1 | 72 |
Gene Symbol: Rxfp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054129.1 | - | 48 |