Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025728.4 | NP_080004.1 | 5, 6 | 753 | ||
NM_029160.3 | NP_083436.2 | 14, 15 | 1594 | |||
GenBank mRNA | AF490391.1 | - | 14, 15 | 1576 | ||
AK014865.1 | - | 5, 6 | 748 | |||
AK016618.1 | - | 5, 6 | 740 |