Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atg10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025770.3 | NP_080046.3 | 4 | 341 | ||
GenBank mRNA | AB079383.1 | - | 3 | 297 | ||
AB091691.1 | - | 3 | 318 | |||
AK009076.1 | - | 4 | 325 | |||
AK017355.1 | - | 4 | 323 | |||
AK077333.1 | - | 4 | 314 | |||
BB843097.1 | - | 4 | 341 | |||
BC023455.1 | - | 3 | 317 | 5249268 |