Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025776.2 | NP_080052.1 | 8 | 810 | ||
GenBank mRNA | AK018452.1 | - | 8 | 833 | ||
AK045116.1 | - | 8 | 830 | |||
AK150271.1 | - | 8 | 823 | |||
AK151758.1 | - | 8 | 823 | |||
AK152252.1 | - | 8 | 823 | |||
AK165277.1 | - | 7 | 2614 | |||
AK166600.1 | - | 8 | 831 | |||
AK167509.1 | - | 8 | 850 | |||
AK168919.1 | - | 8 | 811 | |||
BC042691.1 | - | 8 | 829 | |||
BC080205.1 | - | 8 | 808 | 3709760 |