Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zxdb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081473.1 | NP_001074942.1 | 1 | 2587 | ||
GenBank mRNA | AK016708.1 | - | 1 | 1675 | ||
AK132358.1 | - | 1 | 1730 |
Gene Symbol: Zxda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK155431.1 | - | 2 | 1830 |