Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042407.1 | NP_001035866.1 | 5 | 894 | ||
GenBank mRNA | AK043682.1 | - | 3 | 2263 | ||
BB265326.2 | - | 1 | 148 | |||
BC147304.1 | - | 5 | 879 | 9056372 | ||
BC147305.1 | - | 5 | 879 | 9056374 | ||
BY385428.1 | - | 2 | 254 | |||
CB203398.1 | - | 4 | 468 |