Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smim7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172396.3 | NP_765984.1 | 2, 3 | 76 | ||
GenBank mRNA | AF140607.1 | - | 2, 3 | 76 | ||
AK009731.1 | - | 2, 3 | 86 | |||
AK018610.1 | - | 2, 3 | 89 | |||
AK035786.1 | - | 2, 3 | 77 | |||
AK148562.1 | - | 2, 3 | 88 | |||
AK149156.1 | - | 2, 3 | 88 | |||
AK151539.1 | - | 2, 3 | 77 | |||
AK169625.1 | - | 2, 3 | 88 | |||
BC037511.1 | - | 2, 3 | 57 | 1512313 | ||
BC055692.1 | - | 2, 3 | 77 | 6404961 | ||
BC060963.1 | - | 2, 3 | 94 |