Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133795.1 | NP_598556.1 | 2 | 186 | ||
GenBank mRNA | AK019021.1 | - | 2 | 182 | ||
AK077715.1 | - | 2 | 199 | |||
AK166431.1 | - | 2 | 193 | |||
AK170032.1 | - | 2 | 196 | |||
AK170230.1 | - | 2 | 196 | |||
BC010236.1 | - | 2 | 186 | 3581904 |