Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif26a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001097621.1 | NP_001091090.1 | 12 | 3377 | ||
GenBank mRNA | AK129315.1 | - | 4 | 1501 | ||
BC059062.1 | - | 1 | 208 | |||
BC094358.1 | - | 1 | 882 | |||
BM963109.1 | - | 1 | 78 | |||
GQ489027.1 | - | 12 | 3220 |