Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etfdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025794.2 | NP_080070.2 | 5 | 783 | ||
GenBank mRNA | AK002483.1 | - | 5 | 736 | ||
AK037969.1 | - | 5 | 700 | |||
AK075673.1 | - | 5 | 734 | |||
AK085493.1 | - | 5 | 752 | |||
AK141684.1 | - | 5 | 737 | |||
AK152624.1 | - | 5 | 752 | |||
AK169686.1 | - | 5 | 688 | |||
BC012522.1 | - | 5 | 732 | 2648103 | ||
BC057670.1 | - | 5 | 750 | 2810976 |